Learn More
Cayman Chemical AdenosIn 5-trIphosphateso 1mL
SDP
Supplier:  Cayman Chemical 401821 ML
Encompass_Preferred
A central component of energy storage and metabolism in vivo, providing the metabolic energy to drive metabolic pumps and serving as a coenzyme in a wide array of enzymatic reactions; acts a substrate for kinases involved in cell signaling and of adenylate cyclases that produce the second messenger cAMP
Catalog No. 50-294-2328