Learn More
Cayman Chemical AdenosIn A2A Receptrhumn 10ug
SDP
Catalog No. 503015865
Encompass_Preferred
Source: N-terminal his- and strep-tagged human adenosine A2A receptor containing a TEV protease cleavage site purified from a baculovirus overexpression system in Sf9 cells • MW = ~47.7 kDa
Catalog No. 50-301-5865 Supplier Cayman Chemical Supplier No. 2473010 UG
Source: N-terminal his- and strep-tagged human adenosine A2A receptor containing a TEV protease cleavage site purified from a baculovirus overexpression system in Sf9 cells • MW = ~47.7 kDa