Learn More
Cayman Chemical AdenosIn5OdIphospho 1 700ug
SDP
Catalog No. 502917487
Encompass_Preferred
A nitrophenyl ribose-modified nucleotide and colorimetric PARP1 substrate; hydrolysis releases p-nitrophenol, which can be quantified by colorimetric detection at 405 nm as a measure of PARP1 activity
Catalog No. 50-291-7487 Supplier Cayman Chemical Supplier No. 38357700 UG
A nitrophenyl ribose-modified nucleotide and colorimetric PARP1 substrate; hydrolysis releases p-nitrophenol, which can be quantified by colorimetric detection at 405 nm as a measure of PARP1 activity