Learn More
Cayman Chemical Adenosine-5'-O-diphosphoribose phosphate (sodium salt), Purity: ≥98%, 3.2 mg
SDP
Catalog No. 502916408
Encompass_Preferred
An HMG-CoA reductase inhibitor; selectively inhibits rat liver microsomal HMG-CoA reductase in its non-allosteric form with apparent Ki values of 53 and 550 µM at high and low thiol concentrations, respectively, which are associated with its non-allosteric and allosteric forms
Catalog No. 50-291-6408 Supplier Cayman Chemical Supplier No. 383583.2 MG
An HMG-CoA reductase inhibitor; selectively inhibits rat liver microsomal HMG-CoA reductase in its non-allosteric form with apparent Ki values of 53 and 550 µM at high and low thiol concentrations, respectively, which are associated with its non-allosteric and allosteric forms