Learn More
Cayman Chemical Adenosine-5'-O-triphosphoro-γ-1-(5-sulfonic acid)naphthylamidate (sodium salt), Purity: ≥95%, 3.6 mg
SDP
Catalog No. 502919099
Encompass_Preferred
A fluorescent derivative of ATP; ex/em = 320/460 nm, respectively
Catalog No. 50-291-9099 Supplier Cayman Chemical Supplier No. 385373.6 MG
A fluorescent derivative of ATP; ex/em = 320/460 nm, respectively