Learn More
Medchemexpress LLC Adenosine-1′-13C | 201996-55-6 | 96.3% | 1 MG
SDP
Catalog No. 5000407215
Encompass_Preferred
Adenosine-1′-13C is the 13C labeled Adenosine. Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine affects almost all aspects of cellular physiology and is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07215 Supplier Medchemexpress LLC Supplier No. HYB0228S41MG
Adenosine-1′-13C is the 13C labeled Adenosine. Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine affects almost all aspects of cellular physiology and is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Has potential to affect pharmacokinetic and metabolic profiles of drugs