Learn More
Medchemexpress LLC Adenosine-1′-13C | 201996-55-6 | 96.3% | 5 MG
SDP
Catalog No. 5000406337
Encompass_Preferred
Adenosine-1′-13C is the 13C labeled Adenosine. Adenosine (Adenine riboside) is a ubiquitous endogenous autacoid that acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. It affects almost all aspects of cellular physiology.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06337 Supplier Medchemexpress LLC Supplier No. HYB0228S45MG
Adenosine-1′-13C is the 13C labeled Adenosine. Adenosine (Adenine riboside) is a ubiquitous endogenous autacoid that acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. It affects almost all aspects of cellular physiology.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.