Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine, 1-oxide | 146-92-9 | MFCD00037993 | 99.5% | 283.24 g/mol | C10H13N5O5 | 25 MG
SDP

Catalog No. 5000248414
Encompass_Preferred

Adenosine N1-oxide is a nucleoside derivative used in research as an orally active anti-inflammatory agent. It has been reported to promote osteogenic and adipocyte differentiation and is supplied as a purified research-grade solid with supporting documentation (COA, SDS).

  • CAS 146-92-9
  • Molecular formula C10H13N5O5
  • Molecular weight 283.24 g/mol
  • Purity 99.50%
  • Available in small vial sizes for research use (e.g., 1-25 mg)
  • Documentation: certificate of analysis and safety data sheet available

Catalog No. 50-002-48414 Supplier Medchemexpress LLC Supplier No. HY10208225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine N1-oxide is a nucleoside derivative used in research as an orally active anti-inflammatory agent. It has been reported to promote osteogenic and adipocyte differentiation and is supplied as a purified research-grade solid with supporting documentation (COA, SDS).

  • CAS 146-92-9
  • Molecular formula C10H13N5O5
  • Molecular weight 283.24 g/mol
  • Purity 99.50%
  • Available in small vial sizes for research use (e.g., 1-25 mg)
  • Documentation: certificate of analysis and safety data sheet available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.