Learn More
Medchemexpress LLC Adenosine-13C | 54447-57-3 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0228S51MG
Encompass_Preferred
Adenosine-13C is the 13C labeled Adenosine. Adenosine (Adenine riboside) is a ubiquitous endogenous autacoid that acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. It affects almost all aspects of cellular physiology. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer in analytical methods.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
Catalog No. 50-004-06373