Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine-13C10,15N5 | 202406-75-5 | MFCD01073441 | 99.6% | 282.13 g/mol | 13C10H13 15N5O4 | 10 MG
SDP

Catalog No. 5000297070
Encompass_Preferred

Adenosine-13C10,15N5 is a stable isotope-labeled adenosine supplied as a high-purity analytical standard for use as an internal standard, tracer, or quantitative reference in mass spectrometry and biochemical assays. It contains ten 13C atoms and five 15N atoms and is provided as a solid for laboratory use.

  • High isotopic enrichment and 99.6% purity.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 282.13 g/mol and formula 13C10H13 15N5O4.
  • Soluble in DMSO (33 mg/mL) for solution preparation.
  • Powder stable at -20°C for up to 3 years; in solvent stable at -80°C for 6 months.
  • Supplied in 10 MG quantity for analytical use.

Catalog No. 50-002-97070 Supplier Medchemexpress LLC Supplier No. HYB0228S910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine-13C10,15N5 is a stable isotope-labeled adenosine supplied as a high-purity analytical standard for use as an internal standard, tracer, or quantitative reference in mass spectrometry and biochemical assays. It contains ten 13C atoms and five 15N atoms and is provided as a solid for laboratory use.

  • High isotopic enrichment and 99.6% purity.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 282.13 g/mol and formula 13C10H13 15N5O4.
  • Soluble in DMSO (33 mg/mL) for solution preparation.
  • Powder stable at -20°C for up to 3 years; in solvent stable at -80°C for 6 months.
  • Supplied in 10 MG quantity for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.