Learn More
Medchemexpress LLC Adenosine-13C10,15N5 | 202406-75-5 | MFCD28458760 | >98% | 25 MG
SDP
Catalog No. 5000450983
Encompass_Preferred
Adenosine-13C10,15N5 is a labeled adenosine, an endogenous purine nucleoside that acts as an important homeostatic regulator of physiological function and an anti-arrhythmic agent.
  • Labeled adenosine
  • Endogenous purine nucleoside
  • Acts as an important homeostatic regulator of physiological function
  • Functions as an anti-arrhythmic agent
  • Modulates various physiological processes
  • Activates G protein-coupled receptors (GPCRs), including adenosine receptors (ARs)
  • Involved in energy metabolism
  • Used in research for various diseases
Catalog No. 50-004-50983 Supplier Medchemexpress LLC Supplier No. 25MGHYB0228S9
Adenosine-13C10,15N5 is a labeled adenosine, an endogenous purine nucleoside that acts as an important homeostatic regulator of physiological function and an anti-arrhythmic agent.
  • Labeled adenosine
  • Endogenous purine nucleoside
  • Acts as an important homeostatic regulator of physiological function
  • Functions as an anti-arrhythmic agent
  • Modulates various physiological processes
  • Activates G protein-coupled receptors (GPCRs), including adenosine receptors (ARs)
  • Involved in energy metabolism
  • Used in research for various diseases