Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine-13C5 | 159496-13-6 | 99.9% | 1 MG
SDP

Catalog No. 5000405654
Encompass_Preferred

Adenosine-13C5 is the 13C labeled Adenosine. Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.

  • Isotope-labeled compound, incorporating a stable heavy isotope of carbon (13C).
  • Used as a tracer and internal standard for quantitative analysis by techniques like NMR, GC-MS, or LC-MS.
  • Plays a crucial role in various cellular physiological processes.
  • Purity of 99.9%.
  • Appears as a white to off-white solid.
  • Soluble in DMSO (33.33 mg/mL) and H2O (≥ 6.67 mg/mL).
  • Recommended storage as a powder at -20°C for 3 years or 4°C for 2 years.
  • In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-05654 Supplier Medchemexpress LLC Supplier No. HYB0228S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine-13C5 is the 13C labeled Adenosine. Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.

  • Isotope-labeled compound, incorporating a stable heavy isotope of carbon (13C).
  • Used as a tracer and internal standard for quantitative analysis by techniques like NMR, GC-MS, or LC-MS.
  • Plays a crucial role in various cellular physiological processes.
  • Purity of 99.9%.
  • Appears as a white to off-white solid.
  • Soluble in DMSO (33.33 mg/mL) and H2O (≥ 6.67 mg/mL).
  • Recommended storage as a powder at -20°C for 3 years or 4°C for 2 years.
  • In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.