Learn More
Medchemexpress LLC Adenosine-13C5 | 159496-13-6 | 99.9% | 272.20 | C10H13N5O4 (five 13C atoms) | 10 MG
SDP
Catalog No. 5000297217
Encompass_Preferred
Adenosine-13C5 is a stable isotope-labeled adenosine supplied as a powdered research reagent for use as an internal standard or tracer in mass spectrometry and metabolic studies. It contains five 13C-labeled carbons, has a molecular weight of 272.20, and is typically supplied with high purity for quantitative analysis.
  • Stable isotope-labeled internal standard for quantitative LC-MS/MS.
  • Five 13C labels produce a clear mass shift for tracer studies.
  • High purity (99.9%) supports accurate quantitation.
  • Supplied as a powder suitable for long-term storage under frozen conditions.
  • Available in small mg-scale quantities for analytical applications.
Catalog No. 50-002-97217 Supplier Medchemexpress LLC Supplier No. HYB0228S110MG
Adenosine-13C5 is a stable isotope-labeled adenosine supplied as a powdered research reagent for use as an internal standard or tracer in mass spectrometry and metabolic studies. It contains five 13C-labeled carbons, has a molecular weight of 272.20, and is typically supplied with high purity for quantitative analysis.
  • Stable isotope-labeled internal standard for quantitative LC-MS/MS.
  • Five 13C labels produce a clear mass shift for tracer studies.
  • High purity (99.9%) supports accurate quantitation.
  • Supplied as a powder suitable for long-term storage under frozen conditions.
  • Available in small mg-scale quantities for analytical applications.