Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine-15N5 | 168566-57-2 | 99.9% | 272.21 | C10H13(15N)5O4 | 10 MG
SDP

Catalog No. 5000298266
Encompass_Preferred

Adenosine-15N5 is an isotopically labeled adenosine in which five nitrogen atoms are 15N. It is supplied as a 10 mg solid intended for use as an internal standard, tracer, or reference in mass spectrometry, metabolic studies, and biochemical assays. The material is provided with supporting documentation and handling guidance.

  • Isotopically labeled with five 15N atoms.
  • High purity (99.9%).
  • Molecular weight 272.21 g/mol.
  • Supplied as a 10 mg solid vial.
  • Soluble in DMSO (33.33 mg/mL); ultrasonic and warming recommended.
  • Includes COA, SDS, and handling instructions.
  • Suitable as an internal standard or tracer for analytical applications.

Catalog No. 50-002-98266 Supplier Medchemexpress LLC Supplier No. HYB0228S1110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine-15N5 is an isotopically labeled adenosine in which five nitrogen atoms are 15N. It is supplied as a 10 mg solid intended for use as an internal standard, tracer, or reference in mass spectrometry, metabolic studies, and biochemical assays. The material is provided with supporting documentation and handling guidance.

  • Isotopically labeled with five 15N atoms.
  • High purity (99.9%).
  • Molecular weight 272.21 g/mol.
  • Supplied as a 10 mg solid vial.
  • Soluble in DMSO (33.33 mg/mL); ultrasonic and warming recommended.
  • Includes COA, SDS, and handling instructions.
  • Suitable as an internal standard or tracer for analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.