Learn More
Medchemexpress LLC Adenosine 2',3'-cyclic phosphate sodium | 37063-35-7 | 99.1% | 351.19 | 5 MG
SDP
Catalog No. 5000453623
Encompass_Preferred
Adenosine 2',3'-cyclic phosphate sodium is a 2',3'-cyclic purine nucleotide. It is deaminated by adenosine deaminase and can be degraded to 2'-AMP and 3'-AMP.
  • Efficiently hydrolyzed by the monocations of alkylenediamines at pH 9.0, 50°C.
  • Deaminated by adenosine deaminase from Drosophila larvae.
Catalog No. 50-004-53623 Supplier Medchemexpress LLC Supplier No. HYW0112565MG
Adenosine 2',3'-cyclic phosphate sodium is a 2',3'-cyclic purine nucleotide. It is deaminated by adenosine deaminase and can be degraded to 2'-AMP and 3'-AMP.
  • Efficiently hydrolyzed by the monocations of alkylenediamines at pH 9.0, 50°C.
  • Deaminated by adenosine deaminase from Drosophila larvae.