Learn More
Medchemexpress LLC Adenosine 2',5'-diphosphate sodium | 154146-84-6 | MFCD00009228 | 1 MG
SDP
Catalog No. 5000417702
Encompass_Preferred
Adenosine 2',5'-diphosphate sodium is a potent competitive antagonist of the P2Y1 receptor, and also exhibits non-selective antagonism at recombinant and human platelet P2X1 receptors. This endogenous metabolite is supplied as a white to off-white solid with 99.0% purity.
Key Features & Applications:
  • Competitive P2Y1 antagonist
  • Non-selective P2X1 receptor antagonism in platelets
  • Research use only
Catalog No. 50-004-17702 Supplier Medchemexpress LLC Supplier No. HYN77401MG
Adenosine 2',5'-diphosphate sodium is a potent competitive antagonist of the P2Y1 receptor, and also exhibits non-selective antagonism at recombinant and human platelet P2X1 receptors. This endogenous metabolite is supplied as a white to off-white solid with 99.0% purity.
Key Features & Applications:
  • Competitive P2Y1 antagonist
  • Non-selective P2X1 receptor antagonism in platelets
  • Research use only