Learn More
Medchemexpress LLC Adenosine, 2-amino-2'-deoxy- | 4546-70-7 | MFCD00047240 | 99.6% | 266.26 g·mol⁻¹ | C10H14N6O3 | 5 G
SDP
Catalog No. 5000300586
Encompass_Preferred
2-Amino-2'-deoxyadenosine is a modified deoxyribonucleoside (CAS 4546-70-7) used as a building block in oligonucleotide synthesis and nucleic acid research. Supplied for research use only, it is offered in solid and solution formats and accompanied by analytical documentation to support quality and reproducibility.
  • High purity (99.6%).
  • Available in multiple sizes, including 5 g and solution formats.
  • Molecular weight 266.26 g·mol⁻¹; formula C10H14N6O3.
  • Certificate of analysis available for batch verification.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-003-00586 Supplier Medchemexpress LLC Supplier No. HYW0160415G
2-Amino-2'-deoxyadenosine is a modified deoxyribonucleoside (CAS 4546-70-7) used as a building block in oligonucleotide synthesis and nucleic acid research. Supplied for research use only, it is offered in solid and solution formats and accompanied by analytical documentation to support quality and reproducibility.
  • High purity (99.6%).
  • Available in multiple sizes, including 5 g and solution formats.
  • Molecular weight 266.26 g·mol⁻¹; formula C10H14N6O3.
  • Certificate of analysis available for batch verification.
  • Intended for research use only; not for human or veterinary use.