Learn More
Medchemexpress LLC Adenosine-3',5'-bisphosphate ammonium solution (100 mM) 200 μL
SDP
Catalog No. NC4020192
Encompass_Preferred
Adenosine-3',5'-bisphosphate ammonium is a XRN1 inhibitor (IC50 = 36 nM). Adenosine-3',5'-bisphosphate ammonium can be used in the study of colorectal cancer, breast cancer and lung cancer.
Catalog No. NC4020192 Supplier Medchemexpress LLC Supplier No. HY173593A200UL
Adenosine-3',5'-bisphosphate ammonium is a XRN1 inhibitor (IC50 = 36 nM). Adenosine-3',5'-bisphosphate ammonium can be used in the study of colorectal cancer, breast cancer and lung cancer.