Learn More
Medchemexpress LLC Adenosine 3',5'-cyclic monophosphate | 60-92-4 | MFCD00005845 | 99.9% | 329.21 g/mol | C10H12N5O6P | 10 MG
SDP
Catalog No. 5000297278
Encompass_Preferred
Cyclic adenosine monophosphate (adenosine 3',5'-cyclic monophosphate; cAMP) is provided as an analytical standard for use as a calibration or reference material in quantitative biochemical and analytical assays. The material is supplied in a 10 mg vial and is intended for method development and accurate quantification of cAMP.
  • Analytical standard for quantitative assays.
  • High purity: 99.9% (typical).
  • Suitable for HPLC, LC-MS, and ELISA calibration.
  • Molecular weight 329.21 g/mol; CAS 60-92-4.
Catalog No. 50-002-97278 Supplier Medchemexpress LLC Supplier No. HYB1511R10MG
Cyclic adenosine monophosphate (adenosine 3',5'-cyclic monophosphate; cAMP) is provided as an analytical standard for use as a calibration or reference material in quantitative biochemical and analytical assays. The material is supplied in a 10 mg vial and is intended for method development and accurate quantification of cAMP.
  • Analytical standard for quantitative assays.
  • High purity: 99.9% (typical).
  • Suitable for HPLC, LC-MS, and ELISA calibration.
  • Molecular weight 329.21 g/mol; CAS 60-92-4.