Learn More
Medchemexpress LLC Adenosine 3′-monophosphate | 84-21-9 | MFCD00005746 | 99.0% | 347.22 | C10H14N5O7P | 1 MG
SDP
Catalog No. 5000284735
Encompass_Preferred
Adenosine 3′-monophosphate (3′-AMP) is a nucleotide reagent used in biochemical research. It functions as an agonist of cyclic AMP production, raising intracellular cAMP levels and serving in enzymatic assays such as acid phosphatase determination in human serum. It has been reported to inhibit vascular smooth muscle cell proliferation via A2B receptors and is supplied as a solid for laboratory use.
  • Acts as a cyclic AMP production agonist.
  • Increases intracellular cAMP levels concentration-dependently.
  • Used in assays for acid phosphatase activity in human serum.
  • Reported to inhibit vascular smooth muscle cell proliferation via A2B receptors.
  • Supplied as a white to off-white solid suitable for laboratory handling.
Catalog No. 50-002-84735 Supplier Medchemexpress LLC Supplier No. HYW5874881MG
Adenosine 3′-monophosphate (3′-AMP) is a nucleotide reagent used in biochemical research. It functions as an agonist of cyclic AMP production, raising intracellular cAMP levels and serving in enzymatic assays such as acid phosphatase determination in human serum. It has been reported to inhibit vascular smooth muscle cell proliferation via A2B receptors and is supplied as a solid for laboratory use.
  • Acts as a cyclic AMP production agonist.
  • Increases intracellular cAMP levels concentration-dependently.
  • Used in assays for acid phosphatase activity in human serum.
  • Reported to inhibit vascular smooth muscle cell proliferation via A2B receptors.
  • Supplied as a white to off-white solid suitable for laboratory handling.