Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine, 5′-deoxy-5′-fluoro- | 731-98-6 | 96.2% | 269.23 g/mol | C10H12FN5O3 | 5 MG
SDP

Catalog No. 5000284423
Encompass_Preferred

5′-Fluoro-5′-deoxyadenosine is a fluorinated nucleoside derivative of adenosine supplied for laboratory research use as a biochemical tool and drug derivative. It is intended for research use only and is typically provided in small, milligram quantities for experimental applications.

  • Fluorinated adenosine nucleoside used as a research reagent.
  • CAS number 731-98-6.
  • Molecular formula C10H12FN5O3, molecular weight 269.23 g/mol.
  • Purity 96.22% as supplied.
  • Soluble in DMSO; handling per standard laboratory safety procedures.
  • Available in milligram quantities, including 5 MG.

Catalog No. 50-002-84423 Supplier Medchemexpress LLC Supplier No. HYW6167325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

5′-Fluoro-5′-deoxyadenosine is a fluorinated nucleoside derivative of adenosine supplied for laboratory research use as a biochemical tool and drug derivative. It is intended for research use only and is typically provided in small, milligram quantities for experimental applications.

  • Fluorinated adenosine nucleoside used as a research reagent.
  • CAS number 731-98-6.
  • Molecular formula C10H12FN5O3, molecular weight 269.23 g/mol.
  • Purity 96.22% as supplied.
  • Soluble in DMSO; handling per standard laboratory safety procedures.
  • Available in milligram quantities, including 5 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.