Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenosine 5′-diphosphate sodium salt, 20398-34-9, MFCD00065470, 100 mg
Catalog No. NC0549972
Encompass
Empirical Formula (Hill Notation): C10H15N5O10P2, Molecular Weight: 427.20, bacterial, ≥95% (HPLC), storage temp: −20°C, Synonym: ADP.
Catalog No. NC0549972 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A2754100MG
Add to Cart
Add to Cart
Empirical Formula (Hill Notation): C10H15N5O10P2, Molecular Weight: 427.20, bacterial, ≥95% (HPLC), storage temp: −20°C, Synonym: ADP.