Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine 5'-diphosphate (dicyclohexylammonium) | 102029-87-8 | 99.9% | 1 G
SDP

Catalog No. 5000410171
Encompass_Preferred

Adenosine 5'-diphosphate (dicyclohexylammonium) is supplied as a white to off-white solid with a purity of 99.92% by HPLC. This compound is intended for research applications only and has not been fully validated for medical use.

  • Appearance: white to off-white solid
  • Purity of 99.92% by HPLC
  • 1H NMR spectrum consistent with structure
  • Mass spectrometry consistent with structure
  • Suitable for research applications

Catalog No. 50-004-10171 Supplier Medchemexpress LLC Supplier No. HYD08821G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine 5'-diphosphate (dicyclohexylammonium) is supplied as a white to off-white solid with a purity of 99.92% by HPLC. This compound is intended for research applications only and has not been fully validated for medical use.

  • Appearance: white to off-white solid
  • Purity of 99.92% by HPLC
  • 1H NMR spectrum consistent with structure
  • Mass spectrometry consistent with structure
  • Suitable for research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.