Learn More
Medchemexpress LLC Adenosine 5'-diphosphate (disodium) | 16178-48-6 | 99.9% | 1 MG
SDP
Catalog No. 5000409759
Encompass_Preferred
Adenosine 5'-diphosphate disodium is a nucleoside diphosphate, resulting from ATP dephosphorylation by ATPases. This compound functions as a crucial platelet aggregation agent, playing a significant role in hemostasis and the processes involved in the development and extension of arterial thrombosis.
  • Platelet aggregation agent for hemostasis and the development and extension of arterial thrombosis.
  • Induces human platelets aggregation and [Ca2+]i increasing in platelets, B10 cells, and P2Y1-transfected cells (at 10 μmol/L).
  • Increases cAMP levels in human platelets and in B10 cells (at 5 μmol/L).
Catalog No. 50-004-09759 Supplier Medchemexpress LLC Supplier No. HYD0199S1MG
Adenosine 5'-diphosphate disodium is a nucleoside diphosphate, resulting from ATP dephosphorylation by ATPases. This compound functions as a crucial platelet aggregation agent, playing a significant role in hemostasis and the processes involved in the development and extension of arterial thrombosis.
  • Platelet aggregation agent for hemostasis and the development and extension of arterial thrombosis.
  • Induces human platelets aggregation and [Ca2+]i increasing in platelets, B10 cells, and P2Y1-transfected cells (at 10 μmol/L).
  • Increases cAMP levels in human platelets and in B10 cells (at 5 μmol/L).