Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine 5'-diphosphate disodium | 16178-48-6 | 99.92% | 5 G
SDP

Catalog No. 5000409807
Encompass_Preferred

Adenosine 5'-diphosphate disodium is a nucleoside diphosphate formed from the dephosphorylation of ATP by ATPases. This product functions as a platelet aggregation agent, playing a role in hemostasis and the progression of arterial thrombosis.

  • Nucleoside diphosphate.
  • Product of ATP dephosphorylation by ATPases.
  • Acts as a platelet aggregation agent for hemostasis and the development and extension of arterial thrombosis.
  • Induces human platelet aggregation and an increase in intracellular calcium ([Ca2+]i) in platelets, B10 cells, and P2Y1-transfected cells at a concentration of 10 μmol/L in vitro.
  • Increases cAMP levels in human platelets and in B10 cells at a concentration of 5 μmol/L in vitro.
  • Molecular weight: 471.16
  • Formula: C10H13N5Na2O10P2
  • Appearance: Solid, white to off-white.
  • Initial source: Endogenous metabolite.

Catalog No. 50-004-09807 Supplier Medchemexpress LLC Supplier No. HYD01995G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Adenosine 5'-diphosphate disodium is a nucleoside diphosphate formed from the dephosphorylation of ATP by ATPases. This product functions as a platelet aggregation agent, playing a role in hemostasis and the progression of arterial thrombosis.

  • Nucleoside diphosphate.
  • Product of ATP dephosphorylation by ATPases.
  • Acts as a platelet aggregation agent for hemostasis and the development and extension of arterial thrombosis.
  • Induces human platelet aggregation and an increase in intracellular calcium ([Ca2+]i) in platelets, B10 cells, and P2Y1-transfected cells at a concentration of 10 μmol/L in vitro.
  • Increases cAMP levels in human platelets and in B10 cells at a concentration of 5 μmol/L in vitro.
  • Molecular weight: 471.16
  • Formula: C10H13N5Na2O10P2
  • Appearance: Solid, white to off-white.
  • Initial source: Endogenous metabolite.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.