Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADENOSINE-5'-DIPHOSPHATE, MP Biomedicals

Catalog No. ICN16005305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.1 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16005305 5 g
ICN1600531 0.1 g
ICN16005301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN16005305 Supplier MP Biomedicals, Inc Supplier No. 0216005305
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58-64-0
Molecular Formula C10H15N5O10P2
Molecular Weight (g/mol) 427.203
MDL Number MFCD00066473
InChI Key XTWYTFMLZFPYCI-KQYNXXCUSA-N
Synonym adenosine 5'-diphosphate, adenosine diphosphate, adp, adenosine-5'-diphosphate, adenosine pyrophosphate, adenosine 5'-trihydrogen diphosphate, adenosine-diphosphate, adenosine 5'-pyrophosphate, adp nucleotide, 5'-adenylphosphoric acid
PubChem CID 6022
ChEBI CHEBI:16761
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O

Specifications

Color White
Assay Percent Range >96%
Quantity 5 g
Formula Weight 427.2
Physical Form Solid
Chemical Name or Material Adenosine-5'-Diphosphate, Free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.