Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-Diphosphate, MP Biomedicals™

Catalog No. mp021600535 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021600535 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021600535 Supplier MP Biomedicals, Inc Supplier No. 02160053.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Consists of the pyrophosphate group, the pentose sugar ribose, and the nucleobase adenine. Adenosine-5'-Diphosphate, MP Biomedicals is a nucleoside diphosphate that induces human platelet aggregation and non-competitively blocks the stimulated human plate

ADP (or 5’-ADP) is an ester of pyrophosphoric acid with the nucleosideadenosine. It is the product of ATP dephosphorylation by ATPases and it is converted back to ATP by ATP synthases. ATP is an important energy transfer molecule in cells.

ADP was used as a test compound for studying the endothelium-dependent vascular response in salt sensitive (DS) and salt resistant Dahl rats (DR). The product was used to study the different P2-purinergic receptor subtypes on canine vascular smooth muscle and endothelium.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-64-0
Molecular Formula C10H15N5O10P2
Molecular Weight (g/mol) 427.203
InChI Key XTWYTFMLZFPYCI-KQYNXXCUSA-N
Synonym adenosine 5'-diphosphate, adenosine diphosphate, adp, adenosine-5'-diphosphate, adenosine pyrophosphate, adenosine 5'-trihydrogen diphosphate, adenosine-diphosphate, adenosine 5'-pyrophosphate, adp nucleotide, 5'-adenylphosphoric acid
PubChem CID 6022
ChEBI CHEBI:16761
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O

Specifications

Quantity 500 mg
Solubility Information Soluble in water (50mg/mL).
Formula Weight 427.2
Percent Purity ≥96%
Physical Form Powder
Recommended Storage 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.