Learn More
Medchemexpress LLC Adenosine-5'-diphosphoglucose disodium | 102129-65-7 | 95.0% | 633.31 g·mol⁻¹ | C16H23N5Na2O15P2 | 5 MG
SDP
Catalog No. 5000221923
Encompass_Preferred
ADP-Glucose disodium is the disodium salt of adenosine-5'-diphosphoglucose, a nucleotide-sugar intermediate that serves as the glucose donor in the biosynthesis of starch and other polysaccharides. It is supplied for research use in biochemical and enzymology applications involving glycosyltransferases and starch synthases.
  • Nucleotide-sugar substrate for starch and polysaccharide biosynthesis.
  • Suitable for enzymology and glycosyltransferase assays.
  • Provided as a disodium salt to improve aqueous solubility.
  • Reported purity approximately 95.0% for research use.
  • Available in multiple small pack sizes for laboratory experiments.
Catalog No. 50-002-21923 Supplier Medchemexpress LLC Supplier No. HY1368305MG
ADP-Glucose disodium is the disodium salt of adenosine-5'-diphosphoglucose, a nucleotide-sugar intermediate that serves as the glucose donor in the biosynthesis of starch and other polysaccharides. It is supplied for research use in biochemical and enzymology applications involving glycosyltransferases and starch synthases.
  • Nucleotide-sugar substrate for starch and polysaccharide biosynthesis.
  • Suitable for enzymology and glycosyltransferase assays.
  • Provided as a disodium salt to improve aqueous solubility.
  • Reported purity approximately 95.0% for research use.
  • Available in multiple small pack sizes for laboratory experiments.