Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine-5'-diphosphoglucose disodium | 102129-65-7 | MFCD00056005 | 95.0% | C16H23N5Na2O15P2 | 10MG
SDP

Catalog No. 5000259365
Encompass_Preferred

ADP-Glucose disodium (Adenosine-5'-diphosphoglucose disodium) is a nucleotide sugar used as the glucose donor in biosynthesis of storage and structural polysaccharides such as amylose and amylopectin. Supplied as a white to off-white solid with ≥95.0% purity, it is intended for enzymatic and biochemical assays involving starch synthases and glycosyltransferases.

  • White to off-white solid suitable for biochemical assays.
  • Purity ≥95.0% to support enzymatic reactions.
  • Soluble in water: ≥35 mg/mL (55.27 mM).
  • Molecular weight 633.31 g/mol; formula C16H23N5Na2O15P2.
  • Long-term storage at -20°C; in solution store at -80°C for up to 6 months.
  • Available in small research pack sizes (e.g., 5-100 mg).

Catalog No. 50-002-59365 Supplier Medchemexpress LLC Supplier No. HY13683010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ADP-Glucose disodium (Adenosine-5'-diphosphoglucose disodium) is a nucleotide sugar used as the glucose donor in biosynthesis of storage and structural polysaccharides such as amylose and amylopectin. Supplied as a white to off-white solid with ≥95.0% purity, it is intended for enzymatic and biochemical assays involving starch synthases and glycosyltransferases.

  • White to off-white solid suitable for biochemical assays.
  • Purity ≥95.0% to support enzymatic reactions.
  • Soluble in water: ≥35 mg/mL (55.27 mM).
  • Molecular weight 633.31 g/mol; formula C16H23N5Na2O15P2.
  • Long-term storage at -20°C; in solution store at -80°C for up to 6 months.
  • Available in small research pack sizes (e.g., 5-100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.