Learn More
Medchemexpress LLC Adenosine 5′-diphosphoribose | 20762-30-5 | 98.2% | 559.32 | C15H23N5O14P2 | 25 MG
SDP
Catalog No. 5000212232
Encompass_Preferred
Adenosine 5′-diphosphoribose is an endogenous nucleotide derivative used in biochemical and cell biology research to study ADP-ribosylation, DNA damage responses, and TRPM2 channel activation. Supplied as a light yellow powder with defined solubility and storage recommendations, it is suitable for in vitro assays and biochemical experiments.
  • Biologically active endogenous nucleotide derivative.
  • High purity suitable for biochemical assays (98.2%).
  • Light yellow powder form for convenient handling and storage.
  • Water soluble (175 mg/mL; ultrasonic may be required for dissolution).
  • Stable as a powder at -20°C for long-term storage; in solvent stability is limited.
  • Supplied in small research-scale quantities for laboratory use.
Catalog No. 50-002-12232 Supplier Medchemexpress LLC Supplier No. HY10097325MG
Adenosine 5′-diphosphoribose is an endogenous nucleotide derivative used in biochemical and cell biology research to study ADP-ribosylation, DNA damage responses, and TRPM2 channel activation. Supplied as a light yellow powder with defined solubility and storage recommendations, it is suitable for in vitro assays and biochemical experiments.
  • Biologically active endogenous nucleotide derivative.
  • High purity suitable for biochemical assays (98.2%).
  • Light yellow powder form for convenient handling and storage.
  • Water soluble (175 mg/mL; ultrasonic may be required for dissolution).
  • Stable as a powder at -20°C for long-term storage; in solvent stability is limited.
  • Supplied in small research-scale quantities for laboratory use.