Learn More
Medchemexpress LLC Adenosine 5'-diphosphoribose | 20762-30-5 | 98.2% | 559.32 g/mol | C15H23N5O14P2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100973100MG
Encompass_Preferred
Adenosine 5'-diphosphoribose is an endogenous adenosine diphosphate-ribose provided as a light yellow to yellow powder for laboratory research. It is used as a biochemical reagent and reference standard in studies of ADP-ribosylation, nucleotide signaling, and enzymology, with specified storage and solubility guidelines for reliable handling.
  • High purity suitable for analytical and biochemical assays.
  • Stable as a powder when stored at -20°C for long-term storage.
  • Soluble in water; may require sonication for complete dissolution.
  • Available in multiple pack sizes, including 100 mg for routine experiments.
  • Applicable to enzymology, ADP-ribosylation, and nucleotide signaling studies.
Catalog No. 50-002-11895