Learn More
Medchemexpress LLC Adenosine 5'-diphosphoribose | 20762-30-5 | 98.2% | 559.32 g/mol | C15H23N5O14P2 | 5 MG
SDP
Catalog No. 5000212018
Encompass_Preferred
Adenosine 5'-diphosphoribose is an endogenous nucleotide (ADP ribose) consisting of an adenine base linked to a ribose and two phosphate groups. Supplied as a research-grade solid, it is used in biochemical and cellular assays to investigate ADP-ribosylation signaling, DNA damage response pathways, and calcium-permeable TRPM2 channel activation. The material is provided in assay-friendly quantities with supporting documentation for safe handling.
Catalog No. 50-002-12018 Supplier Medchemexpress LLC Supplier No. HY1009735MG
Adenosine 5'-diphosphoribose is an endogenous nucleotide (ADP ribose) consisting of an adenine base linked to a ribose and two phosphate groups. Supplied as a research-grade solid, it is used in biochemical and cellular assays to investigate ADP-ribosylation signaling, DNA damage response pathways, and calcium-permeable TRPM2 channel activation. The material is provided in assay-friendly quantities with supporting documentation for safe handling.