Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine 5′-diphosphoribose (sodium) | 68414-18-6 | 99.5% | 581.30 g/mol | C15H22N5NaO14P2 | 5 MG
SDP

Catalog No. 5000211896
Encompass_Preferred

Adenosine 5′-diphosphoribose sodium is the sodium salt form of adenosine 5′-diphosphoribose supplied as a high-purity solid for biochemical research. It is provided in small milligram pack sizes and should be stored at low temperatures to maintain stability.

  • High purity (99.5%) suitable for biochemical assays.
  • Sodium salt form, solid, white to light yellow.
  • Molecular weight 581.30 g/mol.
  • Molecular formula C15H22N5NaO14P2.
  • CAS number 68414-18-6.
  • Recommended storage: solid at -20°C; in solution long-term at -80°C.

Catalog No. 50-002-11896 Supplier Medchemexpress LLC Supplier No. HY100973A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine 5′-diphosphoribose sodium is the sodium salt form of adenosine 5′-diphosphoribose supplied as a high-purity solid for biochemical research. It is provided in small milligram pack sizes and should be stored at low temperatures to maintain stability.

  • High purity (99.5%) suitable for biochemical assays.
  • Sodium salt form, solid, white to light yellow.
  • Molecular weight 581.30 g/mol.
  • Molecular formula C15H22N5NaO14P2.
  • CAS number 68414-18-6.
  • Recommended storage: solid at -20°C; in solution long-term at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.