Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine 5'-diphosphoribose sodium | 68414-18-6 | MFCD00136017 | 99.5% | 581.30 g·mol⁻1 | C15H22N5NaO14P2 | 1 ML
SDP

Catalog No. 5000211300
Encompass_Preferred

Adenosine 5′-diphosphoribose sodium is supplied as a ready-to-use aqueous solution (10 mM, 1 mL) for biochemical research. It functions as an ADP-ribose reagent and metabolite standard for studies of NAD+ metabolism and ADP-ribosylation, and is characterized by high reported purity and defined storage recommendations.

  • Ready-to-use 10 mM solution in water, 1 mL.
  • Approximately 99.5% purity.
  • Molecular weight 581.30 g·mol⁻1.
  • CAS number 68414-18-6 for unambiguous identification.
  • Suitable as a biochemical reagent and metabolite standard in NAD+ and ADP-ribosylation research.
  • Storage recommended at -80°C in solution for long-term stability.

Catalog No. 50-002-11300 Supplier Medchemexpress LLC Supplier No. HY100973A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine 5′-diphosphoribose sodium is supplied as a ready-to-use aqueous solution (10 mM, 1 mL) for biochemical research. It functions as an ADP-ribose reagent and metabolite standard for studies of NAD+ metabolism and ADP-ribosylation, and is characterized by high reported purity and defined storage recommendations.

  • Ready-to-use 10 mM solution in water, 1 mL.
  • Approximately 99.5% purity.
  • Molecular weight 581.30 g·mol⁻1.
  • CAS number 68414-18-6 for unambiguous identification.
  • Suitable as a biochemical reagent and metabolite standard in NAD+ and ADP-ribosylation research.
  • Storage recommended at -80°C in solution for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.