Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine 5′-diphosphoribose sodium | 68414-18-6 | MFCD00136017 | 99.5% | 581.30 g/mol | C15H22N5NaO14P2 | 100 MG
SDP

Catalog No. 5000211272
Encompass_Preferred

Adenosine 5′-diphosphoribose sodium is the sodium salt of adenosine 5′-diphosphoribose supplied as a solid for use in biochemical and pharmacological research, including studies of nucleotide signaling and ion channel activation. Provided in a 100 mg quantity, the material is intended for sealed, low-temperature storage to preserve stability.

  • High purity suitable for research applications
  • Sodium salt form of adenosine 5′-diphosphoribose
  • White to light yellow solid appearance
  • Molecular weight 581.30 g/mol
  • CAS number 68414-18-6 for unambiguous identification
  • Store sealed and protected from moisture at low temperatures
  • Suitable for biochemical assays involving nucleotide signaling

Catalog No. 50-002-11272 Supplier Medchemexpress LLC Supplier No. HY100973A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine 5′-diphosphoribose sodium is the sodium salt of adenosine 5′-diphosphoribose supplied as a solid for use in biochemical and pharmacological research, including studies of nucleotide signaling and ion channel activation. Provided in a 100 mg quantity, the material is intended for sealed, low-temperature storage to preserve stability.

  • High purity suitable for research applications
  • Sodium salt form of adenosine 5′-diphosphoribose
  • White to light yellow solid appearance
  • Molecular weight 581.30 g/mol
  • CAS number 68414-18-6 for unambiguous identification
  • Store sealed and protected from moisture at low temperatures
  • Suitable for biochemical assays involving nucleotide signaling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.