Learn More
Medchemexpress LLC Adenosine 5'-diphosphoribose sodium | 68414-18-6 | MFCD00136017 | 99.5% | 581.30 g/mol | C15H22N5NaO14P2 | 25 MG
SDP
Catalog No. 5000211955
Encompass_Preferred
Adenosine 5′-diphosphoribose sodium is a high-purity nucleotide metabolite used in biochemical and cellular research to activate TRPM2 Ca2+-permeable channels and investigate nucleotide-mediated signaling. Supplied as a sodium salt in solid and solution formats, it includes solubility guidance for water and DMSO and storage recommendations for sealed, moisture-free conditions.
  • High purity: 99.5% by HPLC.
  • Soluble in water and DMSO with recommended concentrations for in vitro use.
  • Available as solid (5-100 mg) and 10 mM solution in water.
  • Used to activate TRPM2 channels and study nucleotide signaling pathways.
  • Store sealed, away from moisture; follow recommended temperature guidelines.
Catalog No. 50-002-11955 Supplier Medchemexpress LLC Supplier No. HY100973A25MG
Adenosine 5′-diphosphoribose sodium is a high-purity nucleotide metabolite used in biochemical and cellular research to activate TRPM2 Ca2+-permeable channels and investigate nucleotide-mediated signaling. Supplied as a sodium salt in solid and solution formats, it includes solubility guidance for water and DMSO and storage recommendations for sealed, moisture-free conditions.
  • High purity: 99.5% by HPLC.
  • Soluble in water and DMSO with recommended concentrations for in vitro use.
  • Available as solid (5-100 mg) and 10 mM solution in water.
  • Used to activate TRPM2 channels and study nucleotide signaling pathways.
  • Store sealed, away from moisture; follow recommended temperature guidelines.