Learn More
Medchemexpress LLC Adenosine 5′-diphosphoribose sodium | 68414-18-6 | MFCD00136017 | 99.5% | 581.30 g/mol | C15H22N5NaO14P2 | 50 MG
SDP
Catalog No. 5000211301
Encompass_Preferred
Adenosine 5′-diphosphoribose sodium is a research-grade nucleotide derivative used to investigate ADP-ribosylation and nucleotide metabolism in biochemical and cell biology studies. Supplied as a high-purity solid with guidance for storage and solution preparation, the compound is intended for laboratory research applications and requires appropriate handling and storage to maintain stability.
  • High purity (~99.5%) suitable for analytical and assay use.
  • Solid form for accurate weighing and extended shelf life.
  • Available in small pack sizes to reduce waste for limited experiments.
  • Storage recommendations help preserve stability (store sealed at -20°C).
  • Compatible with aqueous and DMSO stock preparations; filter-sterilize aqueous stocks before use.
Catalog No. 50-002-11301 Supplier Medchemexpress LLC Supplier No. HY100973A50MG
Adenosine 5′-diphosphoribose sodium is a research-grade nucleotide derivative used to investigate ADP-ribosylation and nucleotide metabolism in biochemical and cell biology studies. Supplied as a high-purity solid with guidance for storage and solution preparation, the compound is intended for laboratory research applications and requires appropriate handling and storage to maintain stability.
  • High purity (~99.5%) suitable for analytical and assay use.
  • Solid form for accurate weighing and extended shelf life.
  • Available in small pack sizes to reduce waste for limited experiments.
  • Storage recommendations help preserve stability (store sealed at -20°C).
  • Compatible with aqueous and DMSO stock preparations; filter-sterilize aqueous stocks before use.