Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine 5'-(α,β-methylene)diphosphate triammonium | 00-00-0 | MFCD00042991 | 99.9% | 476.32 g·mol⁻¹ | C11H26N8O9P2 | 5 MG
SDP

Catalog No. 5000262463
Encompass_Preferred

MethADP triammonium (Adenosine 5'-(α,β-methylene)diphosphate triammonium) is a nucleotide analog used as a selective CD73 inhibitor for research into the ATP-adenosine pathway. Supplied as the triammonium salt, it is provided for laboratory research use in solid or solution form.

  • CD73 inhibitor for ATP-adenosine pathway studies.
  • High purity: 99.9%.
  • Molecular weight 476.32 g·mol⁻¹; chemical formula C11H26N8O9P2.
  • Appearance: off-white to light yellow solid.
  • Storage: solid at 4°C; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Available in small solid quantities and as a 10 mM solution.

Catalog No. 50-002-62463 Supplier Medchemexpress LLC Supplier No. HY112502C5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MethADP triammonium (Adenosine 5'-(α,β-methylene)diphosphate triammonium) is a nucleotide analog used as a selective CD73 inhibitor for research into the ATP-adenosine pathway. Supplied as the triammonium salt, it is provided for laboratory research use in solid or solution form.

  • CD73 inhibitor for ATP-adenosine pathway studies.
  • High purity: 99.9%.
  • Molecular weight 476.32 g·mol⁻¹; chemical formula C11H26N8O9P2.
  • Appearance: off-white to light yellow solid.
  • Storage: solid at 4°C; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Available in small solid quantities and as a 10 mM solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.