Learn More
Medchemexpress LLC Adenosine 5'-(α,β-methylene)diphosphate triammonium | 99.9% | 476.32 | 1 ML
SDP
Catalog No. 5000436673
Encompass_Preferred
MethADP (Adenosine 5'-(α,β-methylene)diphosphate) triammonium is a CD73 inhibitor. It can be used for the research of the ATP-adenosine pathway. It inhibits 5'-AMP induced adenosine formation in A498 cells at concentrations of 20 and 100 μmol/L.
  • CD73 inhibitor
  • Used for research of ATP-adenosine pathway
Catalog No. 50-004-36673 Supplier Medchemexpress LLC Supplier No. HY112502C10MM/1ML
MethADP (Adenosine 5'-(α,β-methylene)diphosphate) triammonium is a CD73 inhibitor. It can be used for the research of the ATP-adenosine pathway. It inhibits 5'-AMP induced adenosine formation in A498 cells at concentrations of 20 and 100 μmol/L.
  • CD73 inhibitor
  • Used for research of ATP-adenosine pathway