Learn More
Medchemexpress LLC Adenosine 5'-(α,β-methylene) diphosphate (triammonium) | 99.9% | 476.32 | 25 MG
SDP
Catalog No. 5000438822
Encompass_Preferred
MethADP (Adenosine 5'-(α,β-methylene)diphosphate) triammonium is a CD73 inhibitor that can be used for research of the ATP-adenosine pathway.
  • CD73 inhibitor.
  • Can be used for the research of the ATP-adenosine pathway.
  • Inhibits 5'-AMP induced adenosine formation in A498 cells (in vitro).
Catalog No. 50-004-38822 Supplier Medchemexpress LLC Supplier No. 25MGHY112502C
MethADP (Adenosine 5'-(α,β-methylene)diphosphate) triammonium is a CD73 inhibitor that can be used for research of the ATP-adenosine pathway.
  • CD73 inhibitor.
  • Can be used for the research of the ATP-adenosine pathway.
  • Inhibits 5'-AMP induced adenosine formation in A498 cells (in vitro).