Learn More
Medchemexpress LLC HY-A0181A 500mg Medchemexpress, Adenosine 5'-monophosphate monohydrate CAS:18422-05-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0181A500MG
Encompass_Preferred
Medchemexpress, HY-A0181A 500mg Adenosine 5'-monophosphate monohydrate CAS:18422-05-4 Adenosine 5'-monophosphate monohydrate is an adenosine A1 receptor agonist. Adenosine 5'-monophosphate monohydrate has significant antiviral activity against HSV-1 and HSV-2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2333