Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp AMP [Adenosine-5-Monophosphate, Free Acid], 1 Gram
SDP

Supplier:  Research Products International Corp A112401.0

Encompass_Preferred

  • Substance Name: Adenosine-5-Monophosphate, Free Acid
  • CAS Number: 18422-05-4
  • Molecular Formula: C10H14N5O7P • H2O
  • Molecular Weight: 365.24 g/mol

Catalog No. 50-213-108


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.