Learn More
Medchemexpress LLC Adenosine 5'-monophosphate | 61-19-8 | MFCD00149360 | 99.8% | 347.22 | C10H14N5O7P | 10 MG
SDP
Catalog No. 5000298684
Encompass_Preferred
Adenosine 5'-monophosphate (AMP) is a nucleotide involved in cellular energy transfer and signal transduction. This research-grade biochemical is supplied as a solid or solution and is intended for in vitro research applications; the manufacturer lists high purity and molecular weight of 347.22 g/mol.
  • High purity (99.8%) suitable for research use
  • Defined molecular weight of 347.22 g/mol
  • Available as solid and as DMSO solution for flexible use
  • Commonly used in biochemical assays and receptor pharmacology studies
  • Provided with supporting documentation such as datasheets and COA when available
Catalog No. 50-002-98684 Supplier Medchemexpress LLC Supplier No. HYA0181S110MG
Adenosine 5'-monophosphate (AMP) is a nucleotide involved in cellular energy transfer and signal transduction. This research-grade biochemical is supplied as a solid or solution and is intended for in vitro research applications; the manufacturer lists high purity and molecular weight of 347.22 g/mol.
  • High purity (99.8%) suitable for research use
  • Defined molecular weight of 347.22 g/mol
  • Available as solid and as DMSO solution for flexible use
  • Commonly used in biochemical assays and receptor pharmacology studies
  • Provided with supporting documentation such as datasheets and COA when available