Learn More
Abcam Adenosine 5'-monophosphate (AMP) disodium he x ahydrate, AMPK activator, 5G
Catalog No. 502704580
Encompass_Preferred
MW 391.19 g/mol. Nucleotide involved in the reactions of cellular energy transfers. Acts as an activator of AMP-activated protein kinase (AMPK). Used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5'-nucleotidase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4580 Supplier Abcam Supplier No. AB1465385G
MW 391.19 g/mol. Nucleotide involved in the reactions of cellular energy transfers. Acts as an activator of AMP-activated protein kinase (AMPK). Used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5'-nucleotidase.
The product is subject to the following: Abcam Restricted Use Statement