Learn More
Medchemexpress LLC Adenosine 5'-monophosphate (disodium) | 4578-31-8 | 99.6% | 391.18 | 500 MG
SDP
Catalog No. 5000451710
Encompass_Preferred
Adenosine 5'-monophosphate disodium is an orally active purine nucleotide that participates in ATP metabolism. It is also a ligand for the adenosine 2B receptor. It can activate AMPK in skeletal muscle, ameliorating insulin resistance and impaired glucose metabolism. This product can be used for research on diabetes.
  • Orally active purine nucleotide
  • Participates in ATP metabolism
  • Ligand for adenosine 2B receptor
  • Activates AMPK in skeletal muscle
  • Ameliorates insulin resistance and impaired glucose metabolism
  • Can be used for research of diabetes
Catalog No. 50-004-51710 Supplier Medchemexpress LLC Supplier No. HYW011012500MG
Adenosine 5'-monophosphate disodium is an orally active purine nucleotide that participates in ATP metabolism. It is also a ligand for the adenosine 2B receptor. It can activate AMPK in skeletal muscle, ameliorating insulin resistance and impaired glucose metabolism. This product can be used for research on diabetes.
  • Orally active purine nucleotide
  • Participates in ATP metabolism
  • Ligand for adenosine 2B receptor
  • Activates AMPK in skeletal muscle
  • Ameliorates insulin resistance and impaired glucose metabolism
  • Can be used for research of diabetes