Learn More
Medchemexpress LLC Adenosine 5'-monophosphate disodium | 4578-31-8 | MFCD00037042 | 99.9% | 391.18 | 1 G
SDP
Catalog No. 5000452453
Encompass_Preferred
Adenosine 5'-monophosphate disodium is an orally active purine nucleotide that participates in ATP metabolism. It is also a ligand for the adenosine 2B receptor. This compound can activate AMPK in skeletal muscle, ameliorating insulin resistance and impaired glucose metabolism. It can be used for research on diabetes.
  • Orally active purine nucleotide
  • Participates in ATP metabolism
  • Ligand for the adenosine 2B receptor
  • Activates AMPK in skeletal muscle
  • Ameliorates insulin resistance
  • Improves impaired glucose metabolism
  • Can be used for research on diabetes
Catalog No. 50-004-52453 Supplier Medchemexpress LLC Supplier No. HYW0110121G
Adenosine 5'-monophosphate disodium is an orally active purine nucleotide that participates in ATP metabolism. It is also a ligand for the adenosine 2B receptor. This compound can activate AMPK in skeletal muscle, ameliorating insulin resistance and impaired glucose metabolism. It can be used for research on diabetes.
  • Orally active purine nucleotide
  • Participates in ATP metabolism
  • Ligand for the adenosine 2B receptor
  • Activates AMPK in skeletal muscle
  • Ameliorates insulin resistance
  • Improves impaired glucose metabolism
  • Can be used for research on diabetes