Learn More
Apexbio Technology LLC Adenosine 5'-monophosphate monohydrate 18422-05-4 25mg
SDP
Catalog No. 5000088346
Encompass_Preferred
Adenosine 5 -monophosphate monohydrate (CAS 18422-05-4) is a selective small-molecule agonist targeting the adenosine A1 receptor It is designed to activate the receptor thereby modulating intracellular signaling pathways associated with viral replication inhibition Adenosine 5 -monophosphate monohydrate exerts its biological activity primarily through adenosine A1 receptor activation leading to disruption of key steps in herpes simplex virus type-1 (HSV-1) and type-2 (HSV-2) genome synthesis and propagation In in vitro studies Adenosine 5 -monophosphate monohydrate demonstrates inhibition of HSV viral replication with an IC50 value ranging from 10 20 M Based on these pharmacological properties Adenosine 5 -monophosphate monohydrate holds research potential in antiviral drug development and mechanistic studies related to adenosine receptor-mediated signaling
Catalog No. 50-000-88346 Supplier Apexbio Technology LLC Supplier No. C637625
Adenosine 5 -monophosphate monohydrate (CAS 18422-05-4) is a selective small-molecule agonist targeting the adenosine A1 receptor It is designed to activate the receptor thereby modulating intracellular signaling pathways associated with viral replication inhibition Adenosine 5 -monophosphate monohydrate exerts its biological activity primarily through adenosine A1 receptor activation leading to disruption of key steps in herpes simplex virus type-1 (HSV-1) and type-2 (HSV-2) genome synthesis and propagation In in vitro studies Adenosine 5 -monophosphate monohydrate demonstrates inhibition of HSV viral replication with an IC50 value ranging from 10 20 M Based on these pharmacological properties Adenosine 5 -monophosphate monohydrate holds research potential in antiviral drug development and mechanistic studies related to adenosine receptor-mediated signaling