Learn More
Medchemexpress LLC Adenosine 5'-monophosphate (monohydrate) | 18422-05-4 | 98.9% | C10H16N5O8P | 100 MG
SDP
Catalog No. 5000432644
Encompass_Preferred
Adenosine 5'-monophosphate monohydrate is an analytical standard intended for research and analytical applications. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Used as an analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • For research use only
Catalog No. 50-004-32644 Supplier Medchemexpress LLC Supplier No. HYA0181AR100MG
Adenosine 5'-monophosphate monohydrate is an analytical standard intended for research and analytical applications. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Used as an analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • For research use only