Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine 5'-monophosphate monohydrate | 18422-05-4 | 99.8% | C10H16N5O8P | 250 MG
SDP

Catalog No. 5000358704
Encompass_Preferred

Adenosine 5'-monophosphate monohydrate, also known as 5'-AMP monohydrate, is an adenosine A1 receptor agonist. It has demonstrated significant antiviral activity against both HSV-1 and HSV-2 and is intended for research use only.

  • Functions as an adenosine A1 receptor agonist
  • Exhibits significant antiviral activity against HSV-1 and HSV-2
  • Reduces mRNA expression of TNF-α and IL-6 in RAW264.7 cells
  • Attenuates inflammatory responses by reducing NF-κB p65 recruitment
  • Increases hepatic adenosine levels in mice
  • Improves survival rates and liver health in animal models
  • Chemical formula: C10H16N5O8P
  • Molecular weight: 365.24
  • Appearance: White to off-white solid

Catalog No. 50-003-58704 Supplier Medchemexpress LLC Supplier No. 250MGHYA0181AR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine 5'-monophosphate monohydrate, also known as 5'-AMP monohydrate, is an adenosine A1 receptor agonist. It has demonstrated significant antiviral activity against both HSV-1 and HSV-2 and is intended for research use only.

  • Functions as an adenosine A1 receptor agonist
  • Exhibits significant antiviral activity against HSV-1 and HSV-2
  • Reduces mRNA expression of TNF-α and IL-6 in RAW264.7 cells
  • Attenuates inflammatory responses by reducing NF-κB p65 recruitment
  • Increases hepatic adenosine levels in mice
  • Improves survival rates and liver health in animal models
  • Chemical formula: C10H16N5O8P
  • Molecular weight: 365.24
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.