Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine 5'-monophosphate monohydrate, 99.7%, MP Biomedicals™

Catalog No. ICN10008001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10008001 1 g
ICN10008025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10008001 Supplier MP Biomedicals, Inc Supplier No. 0210008001
Add to Cart
Add to Cart

Used as a monomer in RNA. It is an ester of phosphoric acid and the nucleoside adenosine. AMP consists of a phosphate group, the sugar ribose, and the nucleobase adenine

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18422-05-4
Molecular Formula C10H16N5O8P
Molecular Weight (g/mol) 365.24
MDL Number MFCD00005750,MFCD00149360,MFCD00005750,MFCD00149360
InChI Key ZOEFQKVADUBYKV-MCDZGGTQSA-N
Synonym 5'-Adenylic acid, 5'-AMP
PubChem CID 122130273
IUPAC Name {[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid hydrate
SMILES O.NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 99.70%
Quantity 1 g
Formula Weight 347.2
Physical Form Crystalline Powder
Chemical Name or Material Adenosine 5'-monophosphate monohydrate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.